Wheaton™ Serum Bottles and Vials
Wheaton™ Serum Bottles and Vials
Actual product may vary
Fisherbrand
Wheaton™ Serum Bottles and Vials
These bottles have exceptional wall strength and superior heat and chemical tolerances
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Color
FKS10011422100 mLAmber
FIS06-406C5 mLClear
FIS06-406G50 mLClear
FIS06-402F100 mLAmber
FIS06-406J100 mLClear
FIS06-406K125 mLClear
Catalog number FKS10011422
Price (SGD)
2,548.00
PK
Add to cart
Capacity (Metric):
100 mL
Color:
Amber
Price (SGD)
2,548.00
PK
Add to cart
These bottles have exceptional wall strength and superior heat and chemical tolerances
Ideal for long- and short-term sample storage and vaccine/injectable drug containers.
  • Clear borosilicate glass conforms to USP Type 1 requirements
  • Amber borosilicate glass conforms to USP Type 1 requirements
  • Shrink wrapped partitioned modules reduce breakage
Specifications
Capacity3.38 oz., 100 mL
Capacity (Metric)100 mL
Capacity (English)3.38 oz.
ColorAmber
Diameter (English)2.04 in.
MaterialBorosilicate Glass
Height (Metric)95 mm
Height (English)3.74 in.
Diameter (Metric)52 mm
Unit SizePK