Sterile Polyethylene Snap Caps
Sterile Polyethylene Snap Caps
Actual product may vary
Sterile Polyethylene Snap Caps
For Falcon Round-Bottom Tubes
Have Questions?
Catalog NumberColor
FKS10011021Natural
Catalog number FKS10011021
Price (SGD)
660.00
PK
Add to cart
Color:
Natural
Price (SGD)
660.00
PK
Add to cart
These polyethylene caps feature a patented, two-position fit; loose for aerobic culturing, tight for sealing.
Falcon™ polyethylene dual-position snap caps for 12x75mm tubes offer both vented and fully closed positions. These sterile caps are packaged 500 per bag and 2,000 per case. RNase/DNase-free.
  • Dual position snap cap, offering vented as well as fully closed options
  • Sterilized by gamma irradiation
  • For use with both polystyrene and polypropylene tubes
  • Specifications
    Product TypeSnap Cap
    MaterialPolyethylene
    ColorNatural
    No. per Pack500/Pk.
    No. per Case2000/Cs.
    For Use With (Equipment)Falcon™ 75 L x 12 mm O.D. 5 mL polystyrene and polypropylene round bottom tubes
    Unit SizePK