96 Well Conical (V) Bottom Plate, Non-Treated Surface, Pack of 1
96 Well Conical (V) Bottom Plate, Non-Treated Surface, Pack of 1
Actual product may vary
96 Well Conical (V) Bottom Plate, Non-Treated Surface, Pack of 1
Nunc 96-Well Polystyrene Conical Bottom MicroWell Plates are optimized for robotics, automated readers and liquid handling systems.
Have Questions?
Catalog NumberQuantity
FKS10009572Case of 50
Catalog number FKS10009572
Price (SGD)
836.00
PK
Add to cart
Quantity:
Case of 50
Price (SGD)
836.00
PK
Add to cart
Thermo Scientific Nunc 96-Well Polystyrene Conical Bottom MicroWell Plates are optimized for robotics, automated readers and liquid handling systems. 96-well conical bottom (V) microplates are ideal for sample recovery.
  • Clear polystyrene has excellent optical properties making it ideal for optical measurements
  • Plates are optimized for robotics, plate readers and liquid handlers
  • Customized bar coding is available
Specifications
BarcodeCustom Barcoding Available
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Array8 x 12
Packaging1/Pk., 50/Cs.
Product LineNunc
ColorClear
Product TypeMicroplate
MaterialPolystyrene
No. of Wells96
SterilitySterile
Surface TreatmentNon-treated
Well ShapeConical Bottom (V-well)
No. per Case50
No. per Pack1
QuantityCase of 50
Unit SizePK