Search Thermo Fisher Scientific
| Catalog Number | Length (Metric) | Sterility | Includes |
|---|---|---|---|
| FKS10006021 | 150 mm | Unplugged | |
| FKS10659932 | 150 mm | Non-sterile | Plugged |
| FKS10026041 | 150 mm | Non-sterile | Unplugged |
| FKS10209381 | 230 mm | Non-sterile | Unplugged |