Check Order Status
Zinc Sulfate Heptahydrate (USP/FCC), Fisher Chemical™
Element Search
Structure Search
Zinc Sulfate Heptahydrate (USP/FCC), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FISZ76-3
3 kg
Poly Bottle
Catalog number
FISZ76-3
Price (SGD)
-
Quantity:
3 kg
Packaging:
Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Zinc Sulfate Heptahydrate
Melting Point
100°C
CAS
7446-20-0
ChemAlert Storage Symbol
Gray
Color
White
View more