Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See All Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Tubes
▸
Hybridization Tubes
▸
Hybridization Oven Bottles and Accessories
Actual product may vary
UVP
Hybridization Oven Bottles and Accessories
Have Questions?
Change view
Catalog Number
Product Type
Material
FISUVP98006703
Rocker Tray
Stainless Steel
FISUVP07019401
Hybridization Bottle (15 cm L)
Borosilicate Glass
2 Options
Catalog number
FISUVP98006703
Price (SGD)
1,272.00
Each
Add to cart
Product Type:
Rocker Tray
Material:
Stainless Steel
Price (SGD)
1,272.00
Each
Add to cart
Specifications
For Use With (Application)
DNA, RNA, or protein identification
For Use With (Equipment)
UVP hybridization ovens
Length (Metric)
31.75 cm
Material
Stainless Steel
Product Type
Rocker Tray