Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Task Lighting
▸
Ultraviolet Lamps and Accessories
▸
Blak-Ray™ B-100A UV Lamps
Actual product may vary
UVP
Blak-Ray™ B-100A UV Lamps
High powered lamps offer brilliant irradiance for optimum fluorescence
Have Questions?
Catalog Number
Model
FISUVP95004422
B100A
Catalog number
FISUVP95004422
Price (SGD)
3,721.00
Each
Add to cart
Model:
B100A
Price (SGD)
3,721.00
Each
Add to cart
Features 100 watts of 365nm longwave UV
Manufactured with scratch-resistant, powder painted aluminum lamp head
Lamp head is connected to transformer base by 2.44m (8 ft.) cord
Handle fits into transformer base allowing lamp head to rotate full 360°
95-0044-03:
Includes 20 ft. (6.1m) primary and secondary cords for additional mobility
Specifications
Light Source
UV-A
Voltage
115 V
Wattage
100 W
Wavelength
365 nm
Model
B100A
Packaging Quantity
1 Ea.
Product Type
Longwave UV Lamp