Compact and Handheld UV Lamps
Actual product may vary
UVP
Compact and Handheld UV Lamps
Essential multipurpose UV lamps for laboratory use
Have Questions?
Catalog NumberProduct TypeModelWavelengthWattage
FISUVP95000602Handheld UV LampUVL-56365 nm6 W
Catalog number FISUVP95000602
Price (SGD)
933.00
Each
Product Type:
Handheld UV Lamp
Model:
UVL-56
Wavelength:
365 nm
Wattage:
6 W
Price (SGD)
933.00
Each
Essential multipurpose UV lamps for laboratory use
Choice of 6w handheld or 4w compact UV lamps.
Handheld UV Lamps
  • Uniquely designed with an ergonomic handle
  • Lightweight 6w lamp for easy handling
  • Durable plastic housing
  • Models available with shortwave, longwave UV or a combination shortwave/longwave version
  • Combination shortwave/longwave version comes with a snap-on wavelength selector for switching between wavelengths L x W x H: 14.9 x 3.2 x 2.5 in. (378 x 81 x 64mm)
Compact Lamps
  • Lightweight 4w lamps fit comfortably in the hand
  • Available models include 254nm shortwave, 365nm longwave or a combination shortwave/longwave version
  • Shortwave/longwave version comes with a snap-on wavelength selector for switching between wavelengths L x W x D: 7.8 x 2.8 x 2.1 in. (198 x 71 x 53mm)
Bacterial identification, specimen staining, titration processes, thin layer chromatography, sterilization and visualization of DNA and RNA, and fluorescence of other materials
Specifications
Voltage115 V
Wattage6 W
Wavelength365 nm
Case MaterialPlastic
Dimensions (L x W x H)14.9 x 3.2 x 2.5 in. (378.46 x 81.28 x 63.5 mm)
Electrical Requirements6 W, 115 V, 60 Hz
Height (English)2.5 in.
Height (Metric)63.5 mm
Length (English)14.9 in.
Length (Metric)378.46 mm
Lighting Intensity1000 μW/cm2
ModelUVL-56
Packaging Quantity1 Ea.
Width (English)3.2 in.
Width (Metric)81.28 mm
Product TypeHandheld UV Lamp
Frequency60 Hz
Unit SizeEach