Compact and Handheld UV Lamp Replacement Tubes
Actual product may vary
UVP
Compact and Handheld UV Lamp Replacement Tubes
For use with 4w and 6w UV lamps.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWavelengthWattage
FISUVP34003401365 nm6 W
FISUVP34001301254 nm6 W
FKS11902041254 nm6 W
FKS12862979365 nm6 W
Catalog number FISUVP34003401
Price (SGD)
113.00
Each
Wavelength:
365 nm
Wattage:
6 W
Price (SGD)
113.00
Each
  • For use with 4w and 6w UV lamps
Specifications
Light SourceUV-A
Voltage230 V
Wattage6 W
Wavelength365 nm
Diameter (English)0.62 in.
Diameter (Metric)15.8 mm
For Use With (Equipment)EL-series UV lamp, 3UV™ lamp, UVSL-26P rechargeable UV lamp, handheld UV lamp and mid size UV viewing cabinets
Length (English)9 in.
Length (Metric)228.6 mm
Packaging Quantity1 Ea.
Product TypeUV Lamp Tube
Unit SizeEach