▸
Task Lighting
▸
Ultraviolet Lamps and Accessories
▸
Compact and Handheld UV Lamp Replacement Tubes
Actual product may vary
UVP
Compact and Handheld UV Lamp Replacement Tubes
For use with 4w and 6w UV lamps.
Have Questions?
Catalog Number
Wavelength
Wattage
FISUVP34001301
254 nm
6 W
Catalog number
FISUVP34001301
Price (SGD)
235.00
Each
Add to cart
Wavelength:
254 nm
Wattage:
6 W
Price (SGD)
235.00
Each
Add to cart
For use with 4w and 6w UV lamps
Specifications
Light Source
UV-C
Voltage
115 V
Wattage
6 W
Wavelength
254 nm
For Use With (Application)
For use with Compact 4-Watt UV Lamp
For Use With (Equipment)
Compact 4-Watt UV Lamp
Product Type
UV Lamp Tube