ScintiVerse™ BD Cocktail (Scintanalyzed™), Fisher Chemical
ScintiVerse™ BD Cocktail (Scintanalyzed™), Fisher Chemical
ScintiVerse™ BD Cocktail (Scintanalyzed™), Fisher Chemical
ScintiVerse™ BD Cocktail (Scintanalyzed™), Fisher Chemical
ScintiVerse™ BD Cocktail (Scintanalyzed™), Fisher Chemical
ScintiVerse™ BD Cocktail (Scintanalyzed™), Fisher Chemical
Universal LSC Cocktail for aqueous, nonaqueous and emulsion counting systems. Contains biodegradable linear alkyl aromatic solvent, which can be classified as nonhazardous liquid by local sewerage regulations
Have Questions?
Catalog number FISSX18-4
Price (SGD)
-
Request bulk or custom format
Chemical Identifiers
CAS67774-74-7, 577-11-7, 9016-45-9, 92-71-7, 13280-61-0
Specifications
Chemical Name or MaterialScintiVerse™ BD Cocktail
CAS67774-74-7
ChemAlert Storage SymbolGray
Physical FormLiquid
Background50 CPM max.
View more
  • Universal LSC Cocktail for aqueous, nonaqueous and emulsion counting systems.
  • Contains biodegradable linear alkyl aromatic solvent, which can be classified as a nonhazardous liquid by local sewerage regulations.