Sterivex™ Sterile Pressure-Driven Devices
Sterivex™ Sterile Pressure-Driven Devices
Actual product may vary
MilliporeSigma
Sterivex™ Sterile Pressure-Driven Devices
Minimize contamination risk to filtered solution by direct-dispensing design. Sterivex Sterile Pressure-Driven Devices are ready-to-use units that filter solutions with volumes of 100 to 2000mL. Available in a variety of fittings, pore sizes and membrane types.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterial (Membrane)Pore SizeOutlet FittingNo. per Pack
FISSVHVB1010PVDF0.45 μmFilling bell10/Pk.
FISSVGP01050PES0.22 μmMale nipple50/Pk.
FISSVGPL10RCPES0.22 μmMale luer lok15/Pk.
Catalog number FISSVHVB1010
Price (SGD)
467.00
PK
Add to cart
Material (Membrane):
PVDF
Pore Size:
0.45 μm
Outlet Fitting:
Filling bell
No. per Pack:
10/Pk.
Price (SGD)
467.00
PK
Add to cart
Minimize contamination risk to filtered solution by direct-dispensing design. Sterivex Sterile Pressure-Driven Devices are ready-to-use units that filter solutions with volumes of 100 to 2000mL. Available in a variety of fittings, pore sizes and membrane types.
  • Sterivex filter units work with syringes, peristaltic pumps or pressure vessels. They eliminate the risk of contamination associated with decanting or pipetting.
  • Sterivex units are designed to dispense into virtually any storage container. The devices are available with different outlet connections (see table).
  • Sterivex filter units can process volumes up to 1,000 mL or 2,000 mL, depending on the type of membrane used in the device. A choice of three different membranes is provided.
  • Filter-sterilize volumes up to 1,000 mL or 2,000 mL, using Sterivex™ pressure-driven filter units (capacity depends on the type of membrane used in the device). With three different membrane options and a variety of outlet connection options, Sterivex™ filters can fit virtually any workflow and dispense into nearly any storage container. Sterivex™ filters work with syringes, peristaltic pumps or pressure vessels and eliminate the risk of contamination associated with decanting or pipetting.
Features & Benefits:
  • Works with a variety of pressure devices including syringes, perstaltic pumps and pressure vessels
  • Comes in multiple membrane types to suit your application needs with capacities up to 2 L
Applications:
Sterilization of Aqueous Solutions
Specifications
ColorClear
ConnectorsFemale Luer Lock with upstream vent
DisposableSingle-use
Filter TypeDurapore
Material (Membrane)PVDF
Pore Size0.45 μm
SterilitySterile
Filtration Area10 cm2
Flow Rate≥650 mL/min (at 0.7 bar 25°C)
For Use With (Application)Fast flow andlow protein binding sterilization of aq. Soln.
Max. Inlet Pressure45 psi
No. per Pack10/Pk.
Outlet FittingFilling bell
Outlet StyleFilling bell
Temperature (English) Operating113°F
Temperature (Metric) Operating45°C
TypeBottle Top Filter
Volume (Metric)2 to 20 L
Height (English)3.3 in.
Height (Metric)8.5 cm
Diameter (English)2.12 in.
Diameter (Metric)5.4 cm
Diameter (Metric) Filter5.4 cm
Unit SizePK