SterilElite™ Tabletop Autoclaves
SterilElite™ Tabletop Autoclaves
Actual product may vary
SterilElite™ Tabletop Autoclaves
Combine compact design with high performance, reliability and safety.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Depth (Metric) OutsideVoltage
FISSTE242200624 L660 mm230 V
FISSTE162200816 L580 mm230 V
Catalog number FISSTE2422006
Price (SGD)
-
Capacity (Metric):
24 L
Depth (Metric) Outside:
660 mm
Voltage:
230 V
Combine compact design with high performance, reliability and safety
Intelligent design for your professional choice.
  • Microprocessor control system provides nine fully automatic programs, including add water, sterilization and dry
  • Eight pre-installed sterilization cycles; custom cycles are not an option for the SterilElite16 or 24
  • Two separate water-level check mechanisms: water tank level and chamber water level
  • Auto dry function
  • Pressure door auto-lock device
  • Chamber protected by pressure switch and steam pressure safety valve
  • Vacuum/Pressure Release button cuts off the power supply, exhausts the chamber of pressure, and releases the chamber vacuum simultaneously
Indicator lights:
  • DOOR OPEN - Door has not been closed completely; all functions stop
  • LOW WATER - Water level is too low
  • COMPLETE - Full cycle has run properly and is complete
Specifications
Depth (English) Outside26 in.
Depth (Metric) Outside660 mm
Capacity (English)25.4 qt.
Capacity (Metric)24 L
Depth (Metric) Chamber432 mm
Diameter (English) Chamber10.23 in.
Depth (English) Chamber17.0 in.
Diameter (Metric) Chamber259.8 mm
For Use With (Application)Steam sterilization
Length (English)26.0 in.
Height (English) Exterior17.0 in.
Height (Metric) Exterior440 mm
Length (Metric)660 mm
ModelFront-loading
Product TypeBenchtop Autoclave
Temperature (Metric)121°C to 135°C
Weight (Metric)56 kg
Height (Metric) Chamber228 mm
Height (English) Chamber9 in.
IncludesEight pre-installed cycles
Frequency50/60 Hz
ProgrammabilityAuto Dry
Voltage230 V
Wattage1800 w
Width (English) Exterior21.25 in.
Width (Metric) Exterior540 mm
Dimensions (L x W x H)660 x 540 x 440 mm
Unit SizeEA