Potassium, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical™
Potassium, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical™
Potassium, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical™
Potassium, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical™
Potassium, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical™
Have Questions?
Catalog NumberQuantityPackaging
FISSP351-100100 mLPoly Bottle
Catalog number FISSP351-100
Price (SGD)
-
Quantity:
100 mL
Packaging:
Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPotassium, Reference Standard Solution
Name Note1000ppm ±1%
PackagingPoly Bottle
CAS7447-40-7
ChemAlert Storage SymbolGray
View more