Filling Solution: 4M KCl saturated with AgCl
Product Image
Filling Solution: 4M KCl saturated with AgCl
For single-junction Ag/AgCl electrodes, Calcium Combination ISEs, inner chamber of many double-junction electrodes
Have Questions?
Catalog NumberBoiling Point
FISSP135-500100°C
Catalog number FISSP135-500
Price (SGD)
-
Boiling Point:
100°C
  • 4M KCl electrolyte saturated with AgCl
  • For all single-junction Ag/AgCl electrodes
  • For Calcium Combination ISEs 13-620-536 and 13-620-537
  • For inner chamber of many double-junction electrodes, except accupHast, accupHast R, acciTupH, accuTuph plus
  • In 500mL Poly Bottle
Specifications
Boiling Point100°C
Capacity (English)16.9 oz.
Capacity (Metric)500 mL
ColorColorless
Concentration4M
For Use With (Equipment)Single Junction Ag/AgCl Electrodes, Calcium Combination ISEs, Inner Chamber of Many Double-junction Electrode
Filling Solution TypeSaturated 4 mol/L KCl
Melting Point-10°C
Water SolubleSoluble
Unit SizeEA