Millex™-GP Sterile Syringe Filter, PES, 0.22 μm (Research Use Only)
Millex™-GP Sterile Syringe Filter, PES, 0.22 μm (Research Use Only)
Actual product may vary
MilliporeSigma
Millex™-GP Sterile Syringe Filter, PES, 0.22 μm (Research Use Only)
Ready-to-use syringe-driven units, designed for sterilizing and clarifying sterile solutions
Have Questions?
Catalog NumberDiameter (Metric)Outlet Style
FISSLMPL25SS25 mmMale Luer Lock
Catalog number FISSLMPL25SS
Price (SGD)
890.00
PK
Add to cart
Diameter (Metric):
25 mm
Outlet Style:
Male Luer Lock
Price (SGD)
890.00
PK
Add to cart
Fast filtration with low protein binding
Millex™ syringe filter units with hydrophobic Fluoropore™ or Millipore Express™ PLUS PES membrane are ideal for in-line sterilization of gases and venting sterile containers, and filters with Fluoropore™ membrane can also be used for sterilizing or clarifying organic solutions. There are also specialized filter units to protect hemodialysis transducers from blood and moisture. The 50 mm Millex™ filter units are especially useful for vacuum line protection. All units are bi-directional.
Features & Benefits:
  • Specialized filter units protect hemodialysis transducers from blood and moisture
  • 50 mm Millex™ units ideal for vacuum line protection
  • All Millex™ Hydrophobic PTFE units are bi-directional in flow
Applications: Sterilizing Gases, Vacuum Line Protection, Venting Sterile Containers, Sterilizing and Clarifying Organic Solutions
Specifications
AutoclavableNot Autoclavable
Color CodeClear
DisposableSingle-use
Material (Membrane)PES
Pore Size0.22 μm
SterilitySterile
Filtration Area3.9 cm2
For Use With (Application)Low protein binding; sterilization of tissue culture media, protein solutions or aqueous solutions
Hold up Volume (After Air Purge)<0.1 mL
Housing MaterialEastar™
Inlet StyleFemale Luer Lock
Outlet StyleMale Luer Lock
Pressure Rating5.2 bar (75 psi)
Temperature (Metric) Operating45°C
Volume (Metric)100 mL
Height (Metric)7.1 cm
Diameter (Metric)25 mm
Unit SizePK