Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Chromatography Syringes
▸
Autosampler Syringes
▸
LC Pump Priming Syringe
Actual product may vary
SGE
LC Pump Priming Syringe
For loop fill injection or sample loop flush
Have Questions?
Catalog Number
Volume (Metric)
FISSG008762
5 mL
Catalog number
FISSG008762
Price (SGD)
-
Volume (Metric):
5 mL
Fixed Luer lock
Specifications
For Use With (Application)
Used for loop fill injection or sample loop flush
Product Type
Manual Syringe
Model
5MDF-LL-GT
Quantity
Ea.
Volume (Metric)
5 mL
Needle Point Style
LC