Replaceable Luer Lock Needles: Side Hole
Actual product may vary
Replaceable Luer Lock Needles: Side Hole
Replaceable Luer Lock Needles: Side Hole
Trajan
Replaceable Luer Lock Needles: Side Hole
Replacement needle for 1mL to 100mL Gas Tight Manual Syringes. 50mm needle length.
Have Questions?
Catalog NumberNeedle GaugeDiameter (Metric) InnerDiameter (Metric) Outer
FISSG-03980323 G0.32 mm0.63 mm
Catalog number FISSG-039803
Price (SGD)
-
Needle Gauge:
23 G
Diameter (Metric) Inner:
0.32 mm
Diameter (Metric) Outer:
0.63 mm
  • Replacement luer lock needles
  • For use with 1mL to 100mL gas tight syringes
  • Needle length: 50mm
  • Side Hole tip style
  • ±1% accuracy and reproducibility
Specifications
MaterialStainless Steel
Needle Gauge23 G
Diameter (English) Inner0.012 in.
Diameter (English) Outer0.024 in.
Diameter (Metric) Inner0.32 mm
Diameter (Metric) Outer0.63 mm
For Use With (Application)Gas chromatography
Length (English) Needle2 in.
Product TypeReplacement Needle
Needle Point StyleSide Hole or Dome
Connection TypeLuer Lock
Unit SizePack