Check Order Status
Syringes with Valves
Actual product may vary
SGE
Syringes with Valves
Prefitted with removable needle and valve or Luer Lock valve
Have Questions?
Change view
Catalog Number
Volume (Metric)
FISSG-009472
25 mL
FISSG009670
50 mL
FISSG009770
100 mL
3 Options
Catalog number
FISSG-009472
Price (SGD)
0.00
EA
Add to cart
Volume (Metric):
25 mL
Price (SGD)
0.00
EA
Add to cart
Low syringe carryover
Reduced nonspecific sample interaction
Increased analyte recovery
Ideal for sample storage and transportation or to pressurize sample prior to injection
Material: Borosilicate glass, PTFE plunger (Gas tight)
With Removable Needle
Needle Length: 50mm (2 in.)
Gauge: 23
Outside Diameter: 0.63mm
Inside Diameter: 0.2mm
Cone Tip
Specifications
Autoclavable
Autoclavable
Material
Borosilicate Glass, PTFE (Plunger)
For Use With (Application)
For sample storage and transportation or to pressurize sample prior to injection
Product Type
Syringe
Volume (Metric)
25 mL
Connection Type
Luer Lock, Removable