Check Order Status
FL-70™ Concentrate
Actual product may vary
FL-70™ Concentrate
Powerful alkaline detergent removes dirt, grime, organic residue, coagulated blood, dried gelatin and stopcock grease
Have Questions?
Catalog Number
Volume (English)
FISSF105-1
33.87 oz.
Catalog number
FISSF105-1
Price (SGD)
-
Volume (English):
33.87 oz.
Contains no polyphosphates or chromates to interfere with analysis
Biodegradable
Approx. 10pH
Use 2% solution for normal jobs and 5% for more difficult tasks
Specifications
Form
Liquid
pH
10
Appearance
Yellow
Boiling Point
>100°C
Composition
Tetrasodium Ethylenediamineteraacetate 1.4%, Sodium Oleate 0.5%, Sodium Bicarbonate 0.1%, Sodium Car
Volume (English)
33.87 oz.