accumet™ Electrode Storage Solution
accumet™ Electrode Storage Solution
Actual product may vary
accumet™ Electrode Storage Solution
Have Questions?
Catalog NumberBoiling Point
FISSE40-1100°C
Catalog number FISSE40-1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Boiling Point:
100°C
Solution of potassium hydrogen phthalate—potassium chloride conditions electrodes when not in use
Specifications
Boiling Point100°C
Capacity (English)33.8 oz.
Capacity (Metric)1000 mL
ColorClear
For Use With (Equipment)Electrode
TypeStorage Solution
Water SolubleMiscible
Unit SizeEA