Check Order Status
Buffer Concentrate, pH 10.00 (Certified), Fisher Chemical
Element Search
Structure Search
Buffer Concentrate, pH 10.00 (Certified), Fisher Chemical
Have Questions?
Catalog Number
Quantity
FISSB141-500
500 mL
Catalog number
FISSB141-500
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS
7447-40-7, 1310-73-2, 139-33-3, 7732-18-5, 10043-35-3
Specifications
Chemical Name or Material
Buffer Concentrate, pH 10.01
CAS
7447-40-7
ChemAlert Storage Symbol
Gray
Color
Undesignated
CAS
1310-73-2
View more