Check Order Status
Buffer Solution, pH 1.00 (Certified), Fisher Chemical
Element Search
Structure Search
Buffer Solution, pH 1.00 (Certified), Fisher Chemical
Have Questions?
Catalog Number
Quantity
FISSB140-500
500 mL
Catalog number
FISSB140-500
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS
7447-40-7, 7647-01-0, 50-00-0, 67-56-1, 7732-18-5
Specifications
Chemical Name or Material
Buffer Solution, pH 1.01
CAS
7447-40-7
ChemAlert Storage Symbol
Gray
Melting Point
0°C
Boiling Point
100°C
View more