Buffer Solution, pH 8.00 (Certified), Fisher Chemical™
Buffer Solution, pH 8.00 (Certified), Fisher Chemical™
Buffer Solution, pH 8.00 (Certified), Fisher Chemical™
Buffer Solution, pH 8.00 (Certified), Fisher Chemical™
Buffer Solution, pH 8.00 (Certified), Fisher Chemical™
Buffer Solution, pH 8.00 (Certified), Fisher Chemical™
Potassium Phosphate Monobasic/Sodium Hydroxide
Have Questions?
Catalog NumberQuantityPackaging
FISSB112-500500 mLPoly Bottle
Catalog number FISSB112-500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Poly Bottle
Request bulk or custom format
Chemical Identifiers
CAS1310-73-2, 7778-77-0, 7732-18-5
Specifications
Chemical Name or MaterialBuffer Solution, pH 8.00
CAS1310-73-2
ChemAlert Storage SymbolGray
Health Hazard 2WARNING!
CAS7778-77-0
View more