Check Order Status
Buffer Solution, pH 7.00, Color-Coded Yellow (Certified), Fisher Chemical™
Element Search
Structure Search
Buffer Solution, pH 7.00, Color-Coded Yellow (Certified), Fisher Chemical™
Have Questions?
Change view
Quantity:
4 L
500 mL
Packaging:
Poly Bottle
Catalog number
FISSB107-4
Price (SGD)
-
Quantity:
4 L
Packaging:
Poly Bottle
Request bulk or custom format
Chemical Identifiers
CAS
7778-77-0, 1310-73-2, 7732-18-5, 1934-21-0
Specifications
Absorbance
Pass Test
Chemical Name or Material
Buffer Solution, pH 7.00, Color-Coded Yellow
CAS
7778-77-0
ChemAlert Storage Symbol
Gray
Melting Point
0°C
View more