Block with 20 x 10.5 mm Holes
Block with 20 x 10.5 mm Holes
Actual product may vary
Block with 20 x 10.5 mm Holes
Block with 20 x 10.5 mm holes.
Have Questions?
Catalog NumberNo. of Places
FISSB10520
Catalog number FISSB105
Price (SGD)
-
No. of Places:
20
Blocks are designed to suit the entire range of Ratek™ dry block heaters, but will also fit many other brands of dry block heaters. Block dimensions are 95 mm x 75 mm x 50 mm and made from solid anodised aluminium. The solid construction gives rapid heat transfer and therefore rapid block heat-up when used in a Ratek™ dry block heater. Temperature uniformity is excellent and all blocks are supplied with a thermometer hole for reference.
  • Precision CNC machined aluminium.
  • Chemical resistant black anodised finish.
  • Thermometer reference hole.
  • Will fit many other brands of block heater.
  • Evaporation manifolds available to suit.
Specifications
DescriptionHole Size: Width 10.5 mm, Depth 43.7 mm. 20 Holes.
MaterialAnodized Aluminum
No. of Places20
Dimensions (L x W x H)95 x 75 x 50 mm
Depth (Metric) Well43.7 mm
Diameter (English) Well10.5 mm
Unit SizePK