▸
Burners and Lighters
▸
Lighters
▸
Gas Lighter
Actual product may vary
United Scientific Supplies
Gas Lighter
Hooded Gas Lighter is safe and economical.
Have Questions?
Catalog Number
For Use With (Equipment)
FISS99453
Gas Burner
Catalog number
FISS99453
Price (SGD)
-
For Use With (Equipment):
Gas Burner
Hood traps gas and protects lighting file.
Replacement flints are available (Cat. No. S99442).
Specifications
For Use With (Equipment)
Gas Burner
Length (English)
7 in.
Length (Metric)
177.8 mm
Length (Metric) Blade
18 cm
Material
Metal
For Use With (Application)
Lighting Gas Burners
Product Type
Gas Lighter