Absorbent Liners
Absorbent Liners
Actual product may vary
Absorbent Liners
Economical, highly absorbent bench liners protect surfaces from spills, allowing for safe and easy cleanup.
Have Questions?
Catalog NumberLength (English)
FISS9812850 ft.
Catalog number FISS98128
Price (SGD)
-
Length (English):
50 ft.
Made of highly absorbent three-ply tissue with a polypropylene backing.
  • Economical
  • Waterproof and resistant to most chemicals
  • Absorb spills
  • Retain liquids
  • Soft and durable
  • Cushion surfaces
  • Can be cut to size
Specifications
Product TypeAbsorbent Liner
MaterialPolypropylene
Absorbancy380 mL/m2
Length (English)50 ft.
Length (Metric)1524 cm
Width (English)20 in.
Width (Metric)50.8 cm
Dimensions (L x W)50 ft. x 20 in. (1524 x 50.8cm)
Colorwhite
For Use With (Application)Protects surfaces and absorbs liquids
Thickness3 ply
Unit SizeEA