4-Way Microtube Racks
4-Way Microtube Racks
Actual product may vary
4-Way Microtube Racks
4-Way Microtube Racks
Fisher Science Education
4-Way Microtube Racks
Create as large of a rack as you need by linking together multiple 4-Way Microtube Racks.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
FISS97679Natural
FISS97680Blue
FISS97685Assorted
Catalog number FISS97679
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Color:
Natural
  • 50 mL Tubes, 4 Wells (30 mm dia.), 15 mL Tubes, 12 Wells (17.5 mm dia.), 0.5 mL Tubes, 32 Wells (8.2 mm dia.), 1.5/2.0 mL Tubes, 32 Wells (11.6 mm dia.)
  • Constructed of polypropylene
  • Autoclavable
Specifications
Product TypeMicrotube Rack
ArrayMulti-Array
AutoclavableAutoclavable
ColorNatural
HoldsTubes (1.5, 15, and 50 mL)
MaterialPolypropylene
Positions4
ShapeRectangular
ShelvesNone
StackableYes, When Empty
SterilityNon-sterile
Quantity5 Pk.
Dimensions (L x W x H)17.4 x 9.5 x 5.2 cm (6.8 x 3.7 x 2 in.)
Length (Metric)17.4 cm
Length (English)6.8 in.
Width (Metric)9.5 cm
Width (English)3.7 in.
Height (Metric)5.2 cm
Height (English)2 in.
Unit SizePK