Blood Typing Kits with Simulated Blood
Blood Typing Kits with Simulated Blood
Actual product may vary
Blood Typing Kits with Simulated Blood
Blood Typing Kits with Simulated Blood
Blood Typing Kits with Simulated Blood
Innovating Science
Blood Typing Kits with Simulated Blood
Determine the ABO or ABO/Rh blood type of four unknown samples.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeFor Use With (Application)
FISS96792Blood Type KitSimulated abo/rh blood typing
FISS96792RInnovating Science RefillSimulated abo/rh blood typing
Catalog number FISS96792
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Product Type:
Blood Type Kit
For Use With (Application):
Simulated abo/rh blood typing
Use hands-on activities to understand the nature and importance of antigen-antibody reactions.
  • Provide accurate simulation of the blood typing procedure
  • No toothpicks, stirring or waiting for results
  • Kit contains materials for 10 groups
Kits use simulated blood only.
Specifications
Certifications/Compliance49 CFR 173.4
Product TypeBlood Type Kit
ColorWhite
Grade7 to 14
Age13+ years
For Use With (Application)Simulated abo/rh blood typing
Includes4 simulated blood samples, 1 bottle simulated anti-a serum, 1 bottle simulated anti-b serum, 1bottle simulated anti-rh serum, 40 blood typing trays, Teacher's manual and student study guide copymasters
Quantity1
Class Size10 Lab Groups
Unit SizeEA