Understanding Blood Type Interactions Through Simulated Blood Typing Kit
Understanding Blood Type Interactions Through Simulated Blood Typing Kit
Actual product may vary
Understanding Blood Type Interactions Through Simulated Blood Typing Kit
Use for deeper understanding of the importance of antigen/antibody interactions.
Have Questions?
Catalog NumberProduct Type
FISS96790Simulated Blood Typing Kit
Catalog number FISS96790
Price (SGD)
-
Product Type:
Simulated Blood Typing Kit
Use simulated blood to determine the ABO/Rh blood type of four individuals, one in need of transfusion. The result determines which of three potential donors would provide the best match for the patient in need.
  • Provides simulation of blood typing procedure
  • No toothpicks, stirring or waiting for results
  • Hands-on activities
  • Kit contains materials for 10 groups
Kit uses simulated blood only.
Specifications
Certifications/Compliance49 CFR 173.4
Product TypeSimulated Blood Typing Kit
ColorWhite
Grade7 to 14
Age13+ years
For Use With (Application)For deeper understanding of the importance of antigen/antibody interactions
IncludesTeacher's manual, student guide, 40 blood typing trays, 4 simulated blood samples, donor no. 1, donor no. 2, donor no. 3, patient, 1 bottle simulated anti-a serum, 1 bottle simulated anti-b serum, 1 bottle simulated anti-rh serum
Quantity1
Class Size10 Lab Groups
Unit SizeEA