SP Scienceware™ Biohazard Disposal Can
SP Scienceware™ Biohazard Disposal Can
Actual product may vary
Fisher Catalogue
SP Scienceware™ Biohazard Disposal Can
Ideal for biological/biohazard materials, broken glass, recyclables or other waste.
Have Questions?
Catalog NumberCapacity (English)
FISS967274 gal.
Catalog number FISS96727
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (English):
4 gal.
Autoclave bags (19 × 23 in. [48.3 × 58.4cm]) fit perfectly over a stainless-steel internal wire frame to keep bags open for safe and easy access.
  • Polypropylene construction
  • Puncture resistant
  • Bright red color for high visibility
  • For biohazard disposal
  • Autoclavable at 250°F (121°C)
  • Dimensions: 9 × 12 × 11.5 in. (22.5 × 30.5 × 29cm)
Specifications
Capacity (English)4 gal.
ColorRed
Dimensions (L x W x H)9 x 12 x 11.5 in. (22.5 x 30.5 x 29 cm)
Height (Metric)29 cm
Length (Metric)22.5 cm
Width (Metric)30.5 cm
AutoclavableAutoclavable
For Use With (Application)Biological or biohazard materials, broken glass, recyclables or other waste and sized for benchtop or floor placement
Width (English)12 in.
Length (English)9 in.
Height (English)11.5 in.
MaterialPolypropylene
Product TypeBiohazard Can
Unit SizeEA