Microscope Slide Dispenser
Actual product may vary
Bel-Art
Microscope Slide Dispenser
Sturdy ABS plastic slide dispenser dispenses one slide at a time.
Have Questions?
Catalog number FISS96706
Price (SGD)
-
Sturdy ABS plastic slide dispenser dispenses one slide at a time.
  • Simple knob operation dispenses slides one at a time
  • Refill the dispenser by lifting cover upwards
  • Stores around 72 slides
  • Stores 25 x 75 mm (1 x 3 in.) slides
  • Keeps slides clean and dust-free
Specifications
TypeSlide Storage Cabinet
Slide Capacity72
Depth (English)4.37 in.
Depth (Metric)111 mm
Device Holds72
For Use With (Equipment)Microscope
Height (English)5.51 in.
Height (Metric)140 mm
Width (English)4.88 in.
Width (Metric)124 mm
Unit SizeEach