Forensic Chemistry of Hair Analysis Kit
Forensic Chemistry of Hair Analysis Kit
Actual product may vary
Forensic Chemistry of Hair Analysis Kit
See how scientists use hair to solve a crime.
Have Questions?
Catalog NumberGrade
FISS966437 to 14
Catalog number FISS96643
Price (SGD)
-
Grade:
7 to 14
Determine the origin of hair samples from a crime scene and compare to hair samples from four known suspects.
  • Students will discover the differences between human and animal hair as well as those among different types of human hair
  • Enough materials for 15 tests
Specifications
Product TypeHair Analysis Kit
ColorWhite
Grade7 to 14
Age13 to 20
For Use With (Application)Determine the origin of hair samples from a crime scene and compare to hair samples from four known suspects
IncludesTeacher's manual and student guide, analysis copy-masters, microscope slides, cover slips, deer hair sample, cat hair sample, four human hair samples, 15 forceps, 15 wax pencils
Quantity1
RequiresLight microscopes
Dimensions (L x W x H)9.75 x 6.75 x 3 in.
Class Size15 Lab Groups
Unit SizeEA