Gel Staining Tray
Actual product may vary
United Scientific Supplies
Gel Staining Tray
Gel Staining Tray for staining, fixing, de-staining, and handling fragile electrophoresis gels and membranes.
Have Questions?
Catalog NumberDimensions (L x W x H)
FISS964215 x 5 x 2 in.
Catalog number FISS96421
Price (SGD)
-
Dimensions (L x W x H):
5 x 5 x 2 in.
  • Polypropylene tray and cover resist temperature extremes within a range from -94° to +158°F (-70° to +70°C).
  • LDPE plug provides a leakproof seal and allows easy liquid drainage.
  • Dimensions: 5 x 5 x 2 in. (12.7 x 12.7 x 5.1cm)
Specifications
For Use With (Equipment)Electrophoresis Gels and Membranes
TypeGel Staining Tray
MaterialPolypropylene
Temperature (English) Operating-94°F to +158°F
Temperature (Metric) Operating-70°C to +70°C
Dimensions (L x W x H)5 x 5 x 2 in.
Unit SizeEach