Check Order Status
Scalpel Blades
Actual product may vary
Scalpel Blades
Quality blades, individually packed in aluminum foil.
Have Questions?
Change view
Catalog Number
Size
FISS95938D
# 23
FISS95937A
# 10
FISS95938A
# 20
FISS95938B
# 21
4 Options
Catalog number
FISS95938D
Price (SGD)
-
Size:
# 23
Blades 10 and 11 are suitable for use with handle No. 3 (S95937)
Blades 20 through 23 are suitable for handle No. 4 (S95938)
Specifications
Product Type
Scalpel Blade
Size
# 23
Length (English)
14.96 in.
Length (Metric)
38 cm
For Use With (Equipment)
For Scalpel Handle # 4