VISTA™ Petri Culture Dishes
VISTA™ Petri Culture Dishes
Actual product may vary
VISTA™ Petri Culture Dishes
Flat, clear dishes feature beaded edges for greater mechanical strength.
Have Questions?
Catalog NumberDiameter (Metric)Height (Metric)
FISS95184D142 mm22 mm
Catalog number FISS95184D
Price (SGD)
-
Diameter (Metric):
142 mm
Height (Metric):
22 mm
Bead also provides a means to equally space the side walls of the bottom and cover, reducing the capillary action of the condensed moisture on the sides.
  • Manufactured from soda lime silica glass
  • Unaffected chemically or thermally in routine sterilizations
  • Withstand repeated sterilization — wet or dry
  • Bottoms have a triangular enamel reference point for serial dilutions
  • Covered dish is not airtight
Specifications
ColorClear
MaterialSoda Lime Glass
ShapeRound
EdgesBeaded
Height (Metric)22 mm
Diameter (Metric)142 mm
Unit SizePK