Super Biohazard Disposal Bags
Actual product may vary
Super Biohazard Disposal Bags
Super Biohazard Disposal Bags
Super Biohazard Disposal Bags
Bel-Art
Super Biohazard Disposal Bags
Protects against dangerous tears and leaks. Super Biohazard Disposal Bags feature a sterilization indicator patch that darkens when exposed to steam sterilization for 20 minutes.
Have Questions?
Catalog NumberDimensions (L x W)
FISS90149B23 x 19 in. (58 x 48 cm)
Catalog number FISS90149B
Price (SGD)
-
Dimensions (L x W):
23 x 19 in. (58 x 48 cm)
  • Protect against dangerous tears and leaks
  • Orange-red bags have standard biohazard warning symbol and precautionary procedures in four languages: English, French, German, and Spanish
  • Extra-heavy-duty bags are 2 mil-thick blended polymer
  • Autoclavable to 121°C (250°F)
  • Sterilization indicator patch darkens when exposed to steam sterilization
Specifications
Capacity (Metric)19 L
Closure TypeZip Tie
Capacity (English)5.0 gal.
ColorRed
Dimensions (L x W)23 x 19 in. (58 x 48 cm)
DisposableSingle-use
Length (Metric)58 cm
SterilityNon-sterile
Width (Metric)48 cm
AutoclavableAutoclavable
IncludesStandard biohazard warning symbol and precautionary procedures in four languages: English, French, German and Spanish
For Use With (Application)Extra-strength, high-impact, high-temperature bags reduce autoclave time
Width (English)19 in.
Length (English)23 in.
Thickness2 mil
MaterialPolymer
Product TypeBiohazard Disposal Bag
Unit SizePack