Laboratory Tubing Fittings Assortment Kit
Actual product may vary
Bel-Art
Laboratory Tubing Fittings Assortment Kit
Fittings are for the most common lab connections and compatible with tubing measuring 3.2 to 12.7 mm.
Have Questions?
Catalog NumberMaterial
FISS90143Polypropylene
Catalog number FISS90143
Price (SGD)
-
Material:
Polypropylene
  • Molded of autoclavable FDA grade polypropylene except quick disconnect fittings, which are polyethylene and not autoclavable or FDA grade
  • Kit contains: 6 Straight Tubing Connectors, 7 Stepped Tubing Connectors, 5 Wye (Y) Tubing Connectors, 3 “T” shaped Tubing Connectors, 5 Quick Disconnects and 2 flow control Plastic Tubing Clamps
  • Plastic organizer box has room for adding other fittings and parts
Specifications
AutoclavableAutoclavable
MaterialPolypropylene
For Use With4.7 to 12.7mm
Includes5 straight connectors, 4 stepped fittings, 8 Y fittings, 4 T connectors, 5 quick-disconnect fittings, 2 flow control tubing clamps
ColorNatural
Product TypeFittings Assortment Kit
Unit SizeEach