Covered Lead Rings for Flasks
Covered Lead Rings for Flasks
Actual product may vary
Covered Lead Rings for Flasks
Protect and stabilize expensive glassware with these Vikem™ vinyl-coated lead rings.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (English) InnerDiameter (Metric) InnerFor Use With (Equipment)Color
FISS90139D2.7 in.7 cm1000 to 4000mL Erlenmeyer flasks immersed in water bathsRed
FISS90139B2 in.5.1 cm250 to 1000mL Erlenmeyer flasks immersed in water bathsRed
Catalog number FISS90139D
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Diameter (English) Inner:
2.7 in.
Diameter (Metric) Inner:
7 cm
For Use With (Equipment):
1000 to 4000mL Erlenmeyer flasks immersed in water baths
Color:
Red
  • Stabilize glassware immersed in water baths; reduces floating, breakage and chemical spills
  • Bright red Vikem™ vinyl coating
  • Chemical-resistant
  • Will not scratch flasks
Specifications
Product TypeLead Ring
AutoclavableNot Autoclavable
ColorRed
HoldsFlasks
MaterialVinyl-coated Lead
ShapeRing
ShelvesNone
StackableYes, When Empty
SterilityNon-sterile
For Use With (Equipment)1000 to 4000mL Erlenmeyer flasks immersed in water baths
Length (Metric)114 mm
Width (Metric)114 mm
Height (Metric)43 mm
Diameter (English) Inner2.7 in.
Diameter (Metric) Inner7 cm
Unit SizeEA