Glass Filtration Assembly
Glass Filtration Assembly
Actual product may vary
Fisherbrand
Glass Filtration Assembly
Glass Filtration Assembly for vacuum filtration of tissue culture media, biological fluids, HPCL and other aqueous solutions.
Have Questions?
Catalog number FISS88322
Price (SGD)
-
  • For use when using a vacuum source to filter liquids into a vessel
  • Filter provides an effective filtration of 9.6 cm2
  • Designed to maximise flow and reduce foaming
  • Assembly includes:
    • 300 mL borosilicate glass funnel
    • Highly integrated sealed 47 mm fritted glass support base
    • 1000 mL flask
    • aluminum clamp holds the filter funnel and support base properly
Specifications
ColorClear
DisposableReusable
Filter TypeManifold
For Use With (Equipment)Dispensing Pressure Vessels
MaterialBorosilicate Glass
SterilityNon-sterile
Quantity1 set
TypeFiltration Vacuum
Volume (Metric)300 mL (Vacuum), 1000 mL (Flask)
Length (English)8 in.
Length (Metric)20.3 cm
Width (English)8 in.
Width (Metric)20.3 cm
Height (English)17.25 in.
Height (Metric)43.8 cm
Diameter (English) Inside8 in.
Diameter (Metric) Inside20.32 cm
Unit SizeEA