Search Fisher Scientific
| Catalog Number | Type | Includes | Quantity | Range |
|---|---|---|---|---|
| FISS72417 | Chlorine Test Paper | Color chart, plastic vial | 200 Pk. | 10-50-100-200 ppm |
| FISS72406 | QAC Test Strip | Color chart, plastic waterproof opaque vial, desiccant cap | 100 Pk. | 50-100-200-400 ppm |
Test chlorine from 10 to 200ppm, iodine from 12 to 100ppm and quaternary ammonium compounds from 50-400ppm, or from 200 to 1500ppm