Insta-Test™ Chlorine Test Strips
Insta-Test™ Chlorine Test Strips
Actual product may vary
Fisherbrand
Insta-Test™ Chlorine Test Strips
Test for free and total chlorine with one strip.
Have Questions?
Catalog NumberQuantity
FISS7233525 Pk.
Catalog number FISS72335
Price (SGD)
-
Quantity:
25 Pk.
  • Range and sensitivity is 0, 0.5, 1, 3, 5, 10 ppm.
Specifications
FormatStrips
No. of Tests25
Quantity25 Pk.
Range0, 0.5, 1, 3, 5, 10 ppm
Unit SizeEA