Poxygrid™ Test Tube Racks
Poxygrid™ Test Tube Racks
Actual product may vary
Fisherbrand
Poxygrid™ Test Tube Racks
Chemical (organic solvents, salts, weak acids and weak alkalis) and corrosion-resistant steel wire racks are coated with heat-cured epoxy
Have Questions?
Catalog NumberPositionsHolds
FISS71789108Tubes (13 to 16 mm)
Catalog number FISS71789
Price (SGD)
-
Positions:
108
Holds:
Tubes (13 to 16 mm)
  • Epoxy-coatings are unaffected by organic solvents, salts, weak acids and weak alkalis
  • Three decks and no legs with a slotted grid bottom
  • Square holes hold test tubes securely
  • Reinforced edges
  • Autoclavable, will withstand a temperature range of -223°C (-369°F) to 121°C (250°F)
Specifications
Product TypeTest Tube Rack
Array9 x 12
AutoclavableAutoclavable
ColorBlue
HoldsTubes (13 to 16 mm)
MaterialEpoxy-coated Steel
Positions108
ShapeRectangular
Shelves3 Tiers
StackableYes, When Empty
SterilityNon-sterile
Dimensions (L x W x H)9.5 x 7.4 x 2.5 in. (242 x 189 x 64 mm)
Length (Metric)242 mm
Length (English)9.48 in.
Width (Metric)189 mm
Width (English)7.4 in.
Height (Metric)64 mm
Height (English)2.51 in.
Diameter (Metric) Opening16 mm
Unit SizeEA