Silicone Bulb-Type Safety Pipet Fillers
Silicone Bulb-Type Safety Pipet Fillers
Actual product may vary
Silicone Bulb-Type Safety Pipet Fillers
Chemical-resistant pipet fillers.
Have Questions?
Catalog number FISS68610
Price (SGD)
-
Chemical-resistant pipet fillers.
  • Glass valves improve worker safety by eliminating mouth pipetting and handling of hazardous materials
  • Long-lasting silicone is highly resistant to organic solvents
  • Fit any pipet with O.D. ranging from 0.19 to 0.38 in. (4.5 to 9 mm)
To Use:
  • Fill and dispense pipets with one hand
  • Expel air by squeezing valve '1' above bulb
  • Draw up liquid by pressing valve '2' on stem
  • To release, press valve '3' on side of stem
Specifications
AutoclavableNot Autoclavable
ColorRed
DisposableReusable
For Use WithPipette
Grip TypeRound
MaterialSilicone
Model3 Valve Standard
Number of Channels1
SterilityNon-sterile
TypePipette Filler
Volume (Metric)50 mL
For Use With (Equipment)Pipet
Unit SizeEA