Metal Multiple Tape Dispenser for 1 in. Tape
Product Image
Metal Multiple Tape Dispenser for 1 in. Tape
Holds multiple rolls.
Have Questions?
Catalog number FISS68154
Price (SGD)
-
  • Dispenses multiple rolls of tape to have a variety of colors at hand
  • Each roll can be up to 500 in. (12.7m) long
Specifications
Product TypeMultiple-Roll Tape Dispenser
CapacityMultiple rolls, each roll can be up to 500 in. L
For Use With (Application)Dispenses multiple rolls of tape to have a variety of colors at hand
Unit SizeEA