Disposable Soda Lime Glass Pipets
Disposable Soda Lime Glass Pipets
Actual product may vary
Disposable Soda Lime Glass Pipets
Ideal for transferring samples to sample cups and for drop counting.
Have Questions?
Catalog NumberLength (Metric)
FISS6705115.24 cm
Catalog number FISS67051
Price (SGD)
-
Length (Metric):
15.24 cm
  • Carton top folds back — tilts carton into convenient dispensing position
  • Tip openings range from 0.80 to 1.20mm
  • Top and tip free of sharp edges
  • Can be cotton plugged (plugs not included)
Specifications
Aspirator PlugYes
Diameter (Metric) Outer0.80 to 1.20 mm
MaterialSoda Lime Glass
IncludesUniform bore, free or sharp edge (tip and top)
Length (English)9 in.
Volume (Metric)2 mL
Length (Metric)15.24 cm
TypePasteur Pipet