Absorbent Underpads
Absorbent Underpads
Actual product may vary
Absorbent Underpads
High-absorbency fibers soak up spills.
Have Questions?
Catalog NumberDimensions (L x W)
FISS6701124 x 20 in.
Catalog number FISS67011
Price (SGD)
-
Dimensions (L x W):
24 x 20 in.
  • Quickly absorb liquids and keep spills from spreading
  • Lie flat, hugging the benchtop to prevent sliding
  • Cushioned surface minimizes breakage of delicate labware
  • Polyethylene backing is chemical resistant
  • Spunbond top layer provides durability even when wet
  • Absorb up to 750mL/m2
Specifications
Product TypeAbsorbent Underpad
MaterialPolyethylene
Absorbancy750 mL/m2
Length (English)24 in.
Length (Metric)610 mm
Width (English)20 in.
Width (Metric)510 mm
Dimensions (L x W)24 x 20 in.
For Use With (Application)Quickly absorb liquids and keep spills from spreading
Quantity50 Pack
Unit SizePK