Soft Glass Rods
Soft Glass Rods
Actual product may vary
Soft Glass Rods
With rounded and fire-polished ends.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (English)Diameter (Metric)Length (English)Length (Metric)
FISS634490.19 in.5 mm7.87 in.200 mm
FIS11-380D0.23 in.6 mm9.84 in.250 mm
Catalog number FISS63449
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Diameter (English):
0.19 in.
Diameter (Metric):
5 mm
Length (English):
7.87 in.
Length (Metric):
200 mm
  • Constructed of glass
  • A stirring rod is used to mix chemicals and liquids for laboratory purposes
Specifications
Diameter (English)0.19 in.
IncludesRounded and fire polished ends
Length (English)7.87 in.
Diameter (Metric)5 mm
Length (Metric)200 mm
MaterialGlass
QuantityCase of 1 pack (72 rods/pk.)
Unit SizeEA