PYREX™ Test Tubes
PYREX™ Test Tubes
Actual product may vary
Fisherbrand
PYREX™ Test Tubes
Beaded rim for durability.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)Length (Metric)Capacity (Metric)
FISS6328916 mm150 mm20 mL
FISS6328510 mm75 mm3 mL
Catalog number FISS63289
Price (SGD)
-
Diameter (Metric):
16 mm
Length (Metric):
150 mm
Capacity (Metric):
20 mL
  • Made from special tubing to provide optimum wall thickness
  • Beaded rims are fire-polished
  • All tubes have enamel marking spots except the 0.98 in. (25mm) size
Specifications
Design TypePlain Beaded Rim
Capacity (Metric)20 mL
Length (Metric)150 mm
MaterialBorosilicate Glass
Label TypeMarking Spot
SterilityNon-sterile
GraduatedNo
Diameter (Metric)16 mm
Unit SizePK